Potassium-2-ethylhexanoate hydrate ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39440-06
39440-14
CAAA39440-06EA
88.8
CAD
CAAA39440-06
CAAA39440-14
Potassium-2-ethylhexanoate hydrate ≥95%
Potassium-2-ethylhexanoate hydrate
Purity: 95%
CAS No.: 3164-85-0
Molecular Formula: C8H15KO2.xH2O
Formula Weight: 182.30(anhy)
Physical Form: Lump/semiliquid
Sensitivity: Moisture Sensitive
Odor: Odorless
MDL No.: MFCD00045896
Formula:
C₈H₁₅KO₂·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149471 CAS Number: 3164-85-0 EINECS: 221-625-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...