Potassium selenite

Supplier: Thermo Scientific Chemicals
Danger

39436-14 39436-22
CAAA39436-14EA 199.08 CAD
CAAA39436-14 CAAA39436-22
Potassium selenite
Potassium selenite

CAS No.: 10431-47-7 Molecular Formula: K2SeO3 Formula Weight: 205.16 Physical Form: Powder Density: 2.851 Sensitivity: Hygroscopic Appearance: White Melting Point: 875° dec. UN No.: 2630 MDL No.: MFCD00049663

Formula: K₂O₃Se
Melting Pt: 875 °C (dec.)
Density: 2.851
Storage Temperature: Ambient
MDL Number: MFCD00049663
CAS Number: 10431-47-7
EINECS: 233-909-8
UN: 2630
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR