Potassium tetracyanonickelate(II) hydrate

Supplier: Thermo Scientific Chemicals
Danger

39434-09 39434-18
CAAA39434-09EA 121.34 CAD
CAAA39434-09 CAAA39434-18
Potassium tetracyanonickelate(II) hydrate
Potassium tetracyanonickelate(II) hydrate

Soluble in water


CAS No.: 339527-86-5 Molecular Formula: K2[Ni(CN)4].xH2O Formula Weight: 240.99(anhy) Physical Form: Crystalline Density: 1.875 Melting Point: 100° -H2O Solubility: Soluble in water UN No.: 1588 MDL No.: MFCD00049669

Formula: K₂[Ni(CN)₄].xH₂O
Melting Pt: 100 °C (-H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00049669
CAS Number: 339527-86-5
EINECS: 238-082-7
UN: 1588
ADR: 6.1,II
Merck Index: 14,7684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR