tert-Butylferrocene 98%

Supplier: Thermo Scientific Chemicals

39405-03 39405-06
CAAA39405-03EA 99.86 CAD
CAAA39405-03 CAAA39405-06
tert-Butylferrocene 98%
tert-Butylferrocene

Purity: 98% CAS No.: 1316-98-9 Molecular Formula: C14H18Fe Formula Weight: 242.15 Physical Form: Liquid Density: 1.201 Refractive Index: 1.5790 Boiling Point: 96°/1mm MDL No.: MFCD00058729

Formula: C₁₄H₁₈Fe
Boiling Pt: 96 °C (1 mmHg)
Density: 1,201
Storage Temperature: Ambient
MDL Number: MFCD00058729
CAS Number: 1316-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR