Tricarbonyl(cyclooctatetraene)iron ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39402-02
39402-04
CAAA39402-02EA
320.74
CAD
CAAA39402-02
CAAA39402-04
Tricarbonyl(cyclooctatetraene)iron ≥98%
Tricarbonyl(cyclooctatetraene)iron
sublimes at 50°/0.1mm
Purity: 98%
CAS No.: 12093-05-9
Molecular Formula: C11H8FeO3
Formula Weight: 244.03
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Red-brown
Melting Point: 93-95°
UN No.: 3466
MDL No.: MFCD00001755
Formula:
C₁₁H₈FeO₃ Melting Pt: 93…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001755 CAS Number: 12093-05-9 UN: 3466 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...