Bis(cyclopentadienyl)molybdenum dichloride ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
39393-02
39393-04
CAAA39393-02EA
164.32
CAD
CAAA39393-02
CAAA39393-04
Bis(cyclopentadienyl)molybdenum dichloride ≥99%
Bis(cyclopentadienyl)molybdenum dichloride
Purity: 99%
CAS No.: 12184-22-4
Molecular Formula: C10H10Cl2Mo
Formula Weight: 297.04
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
UN No.: 3261
MDL No.: MFCD00009948
Formula:
C₁₀H₁₀Cl₂Mo MW: 297.032 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009948 CAS Number: 12184-22-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...