Magnesium sulfamate hydrate ≥99%

Supplier: Thermo Scientific Chemicals
Warning

39381-18 39381-30 39381-A1
CAAA39381-18EA 40.13 CAD
CAAA39381-18 CAAA39381-30 CAAA39381-A1
Magnesium sulfamate hydrate ≥99%
Magnesium sulfamate hydrate

Purity: 99% CAS No.: 23221-99-0 Molecular Formula: Mg(SO3NH2)2.xH2O Formula Weight: 216.48(anhy) Physical Form: Powder MDL No.: MFCD00210632

Formula: Mg(SO₃NH₂)₂·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00210632
CAS Number: 13770-91-7
EINECS: 237-398-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR