Bismuth (III) phosphate ≥99%
Supplier: Thermo Scientific Chemicals
39380-18
39380-30
CAAA39380-18EA
169.06
CAD
CAAA39380-18
CAAA39380-30
Bismuth (III) phosphate ≥99%
Bismuth (III) phosphate
Insoluble in water and alkali. Soluble in hydrochloric acid
Purity: 99%
CAS No.: 10049-01-1
Molecular Formula: BiPO4
Formula Weight: 303.95
Physical Form: Powder
Density: 6.32
Odor: Odorless
Melting Point: dec.
Solubility: Insoluble in water and alkali. Soluble in hydrochloric acid
MDL No.: MFCD00049721
Formula:
BiO₄P Melting Pt: Dec. Density: 6.32 Storage Temperature: Ambient |
MDL Number:
MFCD00049721 CAS Number: 10049-01-1 EINECS: 233-161-2 Merck Index: 14,1275 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...