Magnesium salicylate tetrahydrate, tech.
Supplier: Thermo Scientific Chemicals
39379-22
39379-36
CAAA39379-22EA
64.78
CAD
CAAA39379-22
CAAA39379-36
Magnesium salicylate tetrahydrate, tech.
Magnesium salicylate tetrahydrate
Soluble in water and alcohol
CAS No.: 18917-95-8
Molecular Formula: C14H10MgO6.4H2O
Formula Weight: 370.60 (298.60anhy)
Physical Form: Powder
Sensitivity: Air Sensitive
Odor: Odorless
Solubility: Soluble in water and alcohol
MDL No.: MFCD00045815
Formula:
C₁₄H₁₀MgO₆·4H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00045815 CAS Number: 18917-95-8 EINECS: 242-669-3 Merck Index: 14,5672 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...