Magnesium molybdenum oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

39376-18 39376-30
CAAA39376-18EA 183.28 CAD
CAAA39376-18 CAAA39376-30
Magnesium molybdenum oxide ≥99.9% (metals basis)
Magnesium molybdenum oxide

Purity: 99.9% CAS No.: 13767-03-8 Molecular Formula: MgMoO4 Formula Weight: 184.25 Physical Form: -200 Mesh Powder Density: 2.208 Odor: Odorless MDL No.: MFCD00016195

Formula: MgMoO₄
Density: 2,208
Storage Temperature: Ambient
MDL Number: MFCD00016195
CAS Number: 13767-03-8
EINECS: 237-376-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR