Manganese(II) wolframate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

39374-18 39374-30
CAAA39374-18EA 243.32 CAD
CAAA39374-18 CAAA39374-30
Manganese(II) wolframate ≥99.9% (metals basis)
Manganese(II) wolframate

Purity: 99.9% CAS No.: 14177-46-9 Molecular Formula: MnWO4 Formula Weight: 302.79 Physical Form: -200 Mesh Powder Odor: Odorless MDL No.: MFCD00014212

Formula: MnO₄W
Density: 7,23
Storage Temperature: Ambient
MDL Number: MFCD00014212
CAS Number: 14177-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR