Lithium isobutyrate ≥98%
Supplier: Thermo Scientific Chemicals
39372-14
39372-36
CAAA39372-14EA
54.04
CAD
CAAA39372-14
CAAA39372-22
CAAA39372-36
Lithium isobutyrate ≥98%
Lithium isobutyrate
Purity: 98+%
CAS No.: 25179-23-1
Molecular Formula: LiO2CCH(CH3)2
Formula Weight: 94.03
Physical Form: Powder/ lump
Sensitivity: Moisture Sensitive
MDL No.: MFCD00210622
Formula:
LiO₂CCH(CH₃)₂ Storage Temperature: Ambient |
MDL Number:
MFCD00210622 CAS Number: 25179-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...