Lithium periodate dihydrate, reagent

Supplier: Thermo Scientific Chemicals
Danger

39355-18 39355-30
CAAA39355-18EA 105.39 CAD
CAAA39355-18 CAAA39355-30
Lithium periodate dihydrate, reagent
Lithium periodate dihydrate

CAS No.: 21111-84-2 Molecular Formula: LiIO4.2H2O Formula Weight: 233.87 (197.84anhy) Physical Form: Powder Odor: Odorless Melting Point: 370° UN No.: 1479 MDL No.: MFCD00049474

Formula: LiIO₄·2H₂O
MW: 233.87 g/mol (197.84 anhy)
Melting Pt: >370 °C
Storage Temperature: Ambient
MDL Number: MFCD00049474
CAS Number: 21111-84-2
UN: 1479
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR