Aluminium magnesium isopropoxide ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
39320-04
39320-09
39320-18
CAAA39320-04EA
77.1
CAD
CAAA39320-04
CAAA39320-09
CAAA39320-18
Aluminium magnesium isopropoxide ≥99.95% (metals basis)
Aluminium magnesium isopropoxide
Purity: 99.95+%
CAS No.: 69207-83-6
Molecular Formula: Mg[Al(OCH(CH3)2)4]2
Formula Weight: 550.53
Physical Form: Low melt crystalline mass
Sensitivity: Moisture Sensitive
Boiling Point: 114°/0.2mm
UN No.: 3181
MDL No.: MFCD00070426
Formula:
C₂₄H₅₆Al₂MgO₈ Boiling Pt: 114 °C (0.2 mmHg) Melting Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00070426 CAS Number: 69207-83-6 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...