magnesium pyrophosphate

Supplier: Thermo Scientific Chemicals
Warning

39317-18 39317-30 39317-A1
CAAA39317-18EA 102.86 CAD
CAAA39317-18 CAAA39317-30 CAAA39317-A1
magnesium pyrophosphate
magnesium pyrophosphate

CAS No.: 13446-24-7 Molecular Formula: Mg2P2O7 Formula Weight: 222.57 Physical Form: Powder Appearance: White MDL No.: MFCD00049482

Formula: Mg₂O₇P₂
Storage Temperature: Ambient
MDL Number: MFCD00049482
CAS Number: 13446-24-7
EINECS: 236-595-0
Merck Index: 14,5684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR