Magnesium meso-tetraphenylporphine monohydrate

Supplier: Thermo Scientific Chemicals

39316-02 39316-04
CAAA39316-02EA 230.68 CAD
CAAA39316-02 CAAA39316-04
Magnesium meso-tetraphenylporphine monohydrate
Magnesium meso-tetraphenylporphine monohydrate

CAS No.: 14640-21-2 Molecular Formula: C44H28MgN4.H2O Formula Weight: 655.05 (637.04anhy) Physical Form: Crystalline MDL No.: MFCD00049815

Formula: C₄₄H₂₈MgN₄.H₂O
MW: 655.05 g/mol (637.04 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00049815
CAS Number: 14640-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR