Bromopentacarbonylmanganese(I) ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Manganese pentacarbonyl bromide
39310-03
39310-06
CAAA39310-03EA
445.56
CAD
CAAA39310-03
CAAA39310-06
Bromopentacarbonylmanganese(I) ≥98%
Bromopentacarbonylmanganese(I)
Purity: 98%
CAS No.: 14516-54-2
Molecular Formula: C5BrMnO5
Formula Weight: 274.89
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: Orange
UN No.: 3466
MDL No.: MFCD00049806
Formula:
C₅BrMnO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00049806 CAS Number: 14516-54-2 EINECS: 238-522-8 UN: 3466 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...