Manganese(II) phthalocyanine

Supplier: Thermo Scientific Chemicals
Warning

39309-03 39309-06
CAAA39309-03EA 81.84 CAD
CAAA39309-03 CAAA39309-06
Manganese(II) phthalocyanine
Manganese(II) phthalocyanine

CAS No.: 14325-24-7 Molecular Formula: C32H16MnN8 Formula Weight: 567.47 Physical Form: Crystalline MDL No.: MFCD00049821

Formula: C₃₂H₁₆MnN₈
Storage Temperature: Ambient
MDL Number: MFCD00049821
CAS Number: 14325-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR