1,2-Bis(dimethoxyphosphoryl)benzene ≥99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetramethyl 1,2-phenylenebis(phosphonate)

39273-02 39273-04 39273-09
CAAA39273-02EA 60.67 CAD
CAAA39273-02 CAAA39273-04 CAAA39273-09
1,2-Bis(dimethoxyphosphoryl)benzene ≥99%
1,2-Bis(dimethoxyphosphoryl)benzene

Purity: 99% CAS No.: 15104-46-8 Molecular Formula: C10H16O6P2 Formula Weight: 294.18 Physical Form: Crystalline Melting Point: 80-82° UN No.: 3464 MDL No.: MFCD00142461

Formula: C₁₀H₁₆O₆P₂
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00142461
CAS Number: 15104-46-8
UN: 3464
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR