Pentamethylcyclopentadienyliron dicarbonyl dimer
Supplier: Thermo Scientific Chemicals
|
Danger
|
39266-03
39266-06
CAAA39266-03EA
167.48
CAD
CAAA39266-03
CAAA39266-06
Pentamethylcyclopentadienyliron dicarbonyl dimer
Pentamethylcyclopentadienyliron dicarbonyl dimer
CAS No.: 35344-11-7
Molecular Formula: C24H30Fe2O4
Formula Weight: 493.96
Physical Form: Crystalline
Appearance: Red-purple
UN No.: 3466
MDL No.: MFCD00058862
Formula:
C₂₄H₃₀Fe₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00058862 CAS Number: 35344-11-7 UN: 3466 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...