Iron(II) phthalocyanine ≥96%

Supplier: Thermo Scientific Chemicals
Warning

39262-03 39262-06
CAAA39262-03EA 66.83 CAD
CAAA39262-03 CAAA39262-06
Iron(II) phthalocyanine ≥96%
Iron(II) phthalocyanine

Purity: 96% CAS No.: 132-16-1 Molecular Formula: C32H16FeN8 Formula Weight: 568.38 Physical Form: Crystalline MDL No.: MFCD00015953

Formula: C₃₂H₁₆FeN₈
Storage Temperature: Ambient
MDL Number: MFCD00015953
CAS Number: 132-16-1
EINECS: 205-047-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR