Cyclopentadienylmolybdenum tricarbonyl dimer 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
39245-03
39245-06
CAAA39245-03EA
82.79
CAD
CAAA39245-03
CAAA39245-06
Cyclopentadienylmolybdenum tricarbonyl dimer 98%
Cyclopentadienylmolybdenum tricarbonyl dimer
Purity: 98%
CAS No.: 12091-64-4
Molecular Formula: C16H10Mo2O6
Formula Weight: 490.13
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Purple to red
Melting Point: 220° dec.
UN No.: 3466
MDL No.: MFCD00009947
Formula:
C₁₆H₁₀Mo₂O₆ Melting Pt: 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00009947 CAS Number: 12091-64-4 EINECS: 235-156-0 UN: 3466 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...