Bis(pentamethylcyclopentadienyl)nickel(II)
Supplier: Thermo Scientific Chemicals
|
Danger
|
39238-03
39238-06
39238-MD
CAAA39238-03EA
350.76
CAD
CAAA39238-03
CAAA39238-06
CAAA39238-MD
Bis(pentamethylcyclopentadienyl)nickel(II)
Bis(pentamethylcyclopentadienyl)nickel (II)
CAS No.: 74507-63-4
Molecular Formula: C20H30Ni
Formula Weight: 329.17
Physical Form: Crystalline, Packaged in ampoules
Sensitivity: Air and Moisture Sensitive
Formula:
C₂₀H₃₀Ni Storage Temperature: Ambient |
MDL Number:
MFCD00058704 CAS Number: 74507-63-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...