Bis(1,5-cyclooctadiene)nickel(0)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bis(1,5-cyclooctadiene)nickel(0) (Wax encapsulated) (ca. 0.05mmol/capsule), Ni(COD)2
39234-04
39234-09
CAAA39234-04EA
170.64
CAD
CAAA39234-04
CAAA39234-09
Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel (0)
CAS No.: 1295-35-8
Molecular Formula: C16H24Ni
Formula Weight: 275.06
Physical Form: Crystalline
Sensitivity: Air and Light Sensitive
Appearance: Yellow
Odor: Odorless
Melting Point: 60° dec.
UN No.: 1325
MDL No.: MFCD00058902
Formula:
C₁₆H₂₄Ni Melting Pt: 60 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00058902 CAS Number: 1295-35-8 EINECS: 215-072-0 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...