Gadolinium hexaboride ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Gadolinium boride
39225-03
39225-06
CAAA39225-03EA
72.36
CAD
CAAA39225-03
CAAA39225-06
Gadolinium hexaboride ≥99.5% (metals basis)
Gadolinium hexaboride
Purity: 99.5%
CAS No.: 12008-06-9
Molecular Formula: GdB6
Formula Weight: 222.12
Physical Form: -325 Mesh Powder
Odor: Odorless
MDL No.: MFCD00049942
Formula:
B₆Gd MW: 222.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00049942 CAS Number: 12008-06-9 EINECS: 234-530-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...