Bismuth vanadium tetraoxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bismuth vanadium oxide, Bismuth vanadate

39220-14 39220-22
CAAA39220-14EA 206.98 CAD
CAAA39220-14 CAAA39220-22
Bismuth vanadium tetraoxide ≥99.9% (metals basis)
Bismuth vanadium tetraoxide

Insoluble in water. Soluble in acids


Purity: 99.9% CAS No.: 14059-33-7 Molecular Formula: BiVO4 Formula Weight: 323.00 Physical Form: -200 Mesh Powder Appearance: Red to yellow Solubility: Insoluble in water. Soluble in acids UN No.: 3285 MDL No.: MFCD00064836

Formula: BiO₄V
MW: 323.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064836
CAS Number: 14059-33-7
EINECS: 237-898-0
UN: 3285
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR