Cerium zirconium oxide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals

39216-22 39216-36
CAAA39216-22EA 221.2 CAD
CAAA39216-22 CAAA39216-36
Cerium zirconium oxide ≥99.5% (metals basis)
Cerium zirconium oxide

Reacted product. Actual formulation may vary


Purity: 99.5% CAS No.: 53169-24-7 Molecular Formula: CeO2 + ZrO2 Formula Weight: 295.32 Physical Form: -325 Mesh Powder MDL No.: MFCD00168089

Formula: CeO₂.ZrO₂
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00168089
CAS Number: 53169-24-7
EINECS: 258-407-6

Order Now

Specification Test Results

Form -325 Mesh Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR