Cerium(III) phosphate ≥99%
Supplier: Thermo Scientific Chemicals
39215-14
39215-22
39215-36
CAAA39215-14EA
85.48
CAD
CAAA39215-14
CAAA39215-22
CAAA39215-36
Cerium(III) phosphate ≥99%
Cerium(III) phosphate
Purity: 99% min
CAS No.: 13454-71-2
Molecular Formula: CeO4P
Formula Weight: 235.09
Physical Form: Powder
Density: 5.22
Appearance: Dark yellow
Odor: Odorless
MDL No.: MFCD00049424
Formula:
CeO₄P MW: 235.09 g/mol Density: 5.22 Storage Temperature: Ambient |
MDL Number:
MFCD00049424 CAS Number: 13454-71-2 EINECS: 236-637-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...