Cesium titanium oxide ≥99.85% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

39207-06 39207-14
CAAA39207-06EA 103.02 CAD
CAAA39207-06 CAAA39207-14
Cesium titanium oxide ≥99.85% (metals basis)
Cesium titanium oxide

Purity: 99.85% CAS No.: 12158-57-5 Molecular Formula: Cs2O3Ti Formula Weight: 361.71 Physical Form: -100 Mesh Powder Appearance: White MDL No.: MFCD00053969

Formula: Cs₂O₃Ti
Storage Temperature: Ambient
MDL Number: MFCD00053969
CAS Number: 12158-57-5
EINECS: 235-283-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR