Cesium wolframate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Cesium tungsten oxide, Cesium tungstate

39206-06 39206-14
CAAA39206-06EA 145.83 CAD
CAAA39206-06 CAAA39206-14
Cesium wolframate ≥99.9% (metals basis)
Cesium wolframate

Purity: 99.9% CAS No.: 13587-19-4 Molecular Formula: Cs2O4W Formula Weight: 513.66 Physical Form: -200 Mesh Powder Sensitivity: Hygroscopic Appearance: White to off-white Odor: Odorless MDL No.: MFCD00049438

Formula: Cs₂O₄W
MW: 513.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049438
CAS Number: 13587-19-4
EINECS: 237-019-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR