Zirconium(IV) 2-ethylhexanoate ≥97%

Supplier: Thermo Scientific Chemicals
Warning

39174-14 39174-22
CAAA39174-14EA 119.61 CAD
CAAA39174-14 CAAA39174-22
Zirconium(IV) 2-ethylhexanoate ≥97%
Zirconium(IV) 2-ethylhexanoate

Purity: 97% CAS No.: 2233-42-3 Molecular Formula: C32H60O8Zr Formula Weight: 664.04 Flash Point: 93°(199°F) Physical Form: Viscous liquid Sensitivity: Moisture Sensitive Appearance: Amber MDL No.: MFCD00072684

Formula: C₃₂H₆₀O₈Zr
MW: 664.048 g/mol
Flash Pt: >93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00072684
CAS Number: 2233-42-3
EINECS: 218-776-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR