Tin(IV) 2-ethylhexanoate diisopropoxide ≥98%

Supplier: Thermo Scientific Chemicals
Warning

39169-03 39169-06
CAAA39169-03EA 51.82 CAD
CAAA39169-03 CAAA39169-06 CAAA39169-14
Tin(IV) 2-ethylhexanoate diisopropoxide ≥98%
Tin(IV) 2-ethylhexanoate diisopropoxide

Purity: 98% CAS No.: 288374-32-3 Molecular Formula: C22H44O6Sn Formula Weight: 523.08 Physical Form: Liquid Sensitivity: Moisture Sensitive MDL No.: MFCD00210604

Formula: C₂₂H₄₄O₆Sn
MW: 523.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210604
CAS Number: 288374-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR