Potassium-2-ethylhexanoate solution ≥75% (w/w)

Supplier: Thermo Scientific Chemicals
Warning

39163-04 39163-18
CAAA39163-04EA 58.62 CAD
CAAA39163-04 CAAA39163-18
Potassium-2-ethylhexanoate solution ≥75% (w/w)
Potassium-2-ethylhexanoate

Contains a mixture of organic solvents


Purity: 99.9% CAS No.: 3164-85-0 Molecular Formula: C8H15KO2 Formula Weight: 182.30 Physical Form: Viscous liquid Odor: Characteristic UN No.: 1993 MDL No.: MFCD00045896

Formula: C₈H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00045896
CAS Number: 3164-85-0
EINECS: 221-625-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR