Potassium isopropoxide 5% (w/v) in isopropanol (IPA) , 99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
39162-AC
39162-AE
CAAA39162-ACEA
138.57
CAD
CAAA39162-AC
CAAA39162-AE
Potassium isopropoxide 5% (w/v) in isopropanol (IPA) , 99% (metals basis)
Potassium isopropoxide
Purity: 99%
CAS No.: 6831-82-9
Molecular Formula: C3H7KO
Formula Weight: 98.19
Flash Point: 12°(54°F)
Physical Form: Liquid
Sensitivity: Air and Moisture Sensitive
Appearance: Colorless to pale yellow
Odor: Alcohol-like
UN No.: 3274
MDL No.: MFCD00210641
Formula:
C₃H₇KO MW: 98.186 g/mol Flash Pt: 12 °C (54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00210641 CAS Number: 6831-82-9 EINECS: 229-909-2 UN: 3274 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...