Lithium niobium ethoxide 5% (w/v) in ethanol , 99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
39155-AC
39155-AE
CAAA39155-ACEA
170.64
CAD
CAAA39155-AC
CAAA39155-AE
Lithium niobium ethoxide 5% (w/v) in ethanol , 99% (metals basis)
Lithium niobium ethoxide
Purity: 99+%
CAS No.: 86745-52-0
Molecular Formula: LiNb(OC2H5)6
Formula Weight: 370.21
Flash Point: 17°(63°F)
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Yellow
Odor: Alcohol-like
UN No.: 1170
MDL No.: MFCD00210625
Formula:
LiNb(OC₂H₅)₆ Flash Pt: 17 °C (63 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00210625 CAS Number: 86745-52-0 UN: 1170 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...