Lithium niobium methoxide 5% (w/v) in methanol , 99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
39154-AC
39154-AE
CAAA39154-ACEA
125.14
CAD
CAAA39154-AC
CAAA39154-AE
Lithium niobium methoxide 5% (w/v) in methanol , 99% (metals basis)
Lithium niobium methoxide
Purity: 99+%
CAS No.: 21864-11-9
Molecular Formula: C6H18LiNbO6
Formula Weight: 285.92
Physical Form: Liquid
Sensitivity: Moisture Sensitive
UN No.: 1230
MDL No.: MFCD00210627
Formula:
C₆H₁₈LiNbO₆ MW: 285.92 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210627 CAS Number: 21864-11-9 UN: 1230 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...