Lead(II) titanium(IV) isopropoxide 5% (w/v) in isopropanol (IPA) , 99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
39153-AC
39153-AE
CAAA39153-ACEA
153.1
CAD
CAAA39153-AC
CAAA39153-AE
Lead(II) titanium(IV) isopropoxide 5% (w/v) in isopropanol (IPA) , 99.9% (metals basis)
Lead(II) titanium(IV) isopropoxide
Purity: 99.9%
Molecular Formula: PbTi(OC3H7)6
Formula Weight: 609.29
Flash Point: 12°(54°F)
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown
Odor: Alcohol-like
UN No.: 1219
MDL No.: MFCD00210621
Formula:
PbTi(OC₃H₇)₆ Flash Pt: 12 °C (54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00210621 UN: 1219 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...