Barium niobium oxide 99,9
Supplier: Thermo Scientific Chemicals
|
Warning
|
39130-09
39130-18
CAAA39130-09EA
92.9
CAD
CAAA39130-09
CAAA39130-18
Barium niobium oxide 99,9
Barium niobium oxide
Insoluble in water
Purity: 99.9%
CAS No.: 12009-14-2
Molecular Formula: BaNb2O6
Formula Weight: 419.11
Physical Form: -100 Mesh Powder
Solubility: Insoluble in water
UN No.: 1564
MDL No.: MFCD00064751
Formula:
BaNb₂O₆ MW: 419.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064751 CAS Number: 12009-14-2 EINECS: 234-543-1 UN: 1564 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...