Aluminium stearate, tech.
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum stearate, Aluminum tristearate
39125-A1
39125-A7
CAAA39125-A1EA
95.75
CAD
CAAA39125-A1
CAAA39125-A7
Aluminium stearate, tech.
Aluminium stearate
CAS No.: 637-12-7
Molecular Formula: Al[CH3(CH2)16CO2]3
Formula Weight: 877.42
Physical Form: Powder
Melting Point: 105-115°
Merck Reference: 14,364
MDL No.: MFCD00036389
Formula:
C₅₄H₁₀₅AlO₆ MW: 877.389 g/mol Melting Pt: 105…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036389 CAS Number: 637-12-7 EINECS: 211-279-5 Merck Index: 14,364 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...