Diamond (natural) ≥99.9% (metals basis), powder 40-60 micron

Supplier: Thermo Scientific Chemicals

38984-03 38984-06
CAAA38984-03EA 167.48 CAD
CAAA38984-03 CAAA38984-06
Diamond (natural) ≥99.9% (metals basis), powder 40-60 micron
Diamond

Applications: Diamond powder is generally used as an abrasive to grind and polish hard surfaces.


Purity: 99.9% Physical Form: Powder MDL No.: MFCD00211867


Applications: Diamond powder is generally used as an abrasive to grind and polish hard surfaces.

Storage Temperature: Ambient
MDL Number: MFCD00211867
CAS Number: 7782-40-3
EINECS: 231-953-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR