Tropaeolin O

Supplier: Thermo Scientific Chemicals

38708-04 38708-09
CAAAA38708-04EA 36.18 CAD
CAAAA38708-04 CAAA38708-09
Tropaeolin O
Tropaeolin O

Transition interval: pH 11.0 (yellow) to 12.7 (orange)


CAS No.: 547-57-9 Molecular Formula: C12H9N2NaO5S Formula Weight: 316.27 Physical Form: Powder Appearance: Orange to brown Solubility: Soluble in water and alcohol MDL No.: MFCD00007499

Formula: C₁₂H₉N₂O₅SNa
Storage Temperature: Ambient
MDL Number: MFCD00007499
CAS Number: 547-57-9
EINECS: 208-924-8
Merck Index: 14,9775

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR