Manganese(II) 2-ethylhexanoate 40% (w/w) in mineral spirits (6% Mn)
Supplier: Thermo Scientific Chemicals
|
Danger
|
38607-30
38607-A7
CAAA38607-30EA
49.77
CAD
CAAA38607-30
CAAA38607-A7
Manganese(II) 2-ethylhexanoate 40% (w/w) in mineral spirits (6% Mn)
Manganese(II) 2-ethylhexanoate
CAS No.: 13434-24-7
Molecular Formula: C16H30MnO4
Formula Weight: 341.35
Flash Point: 40°(104°F)
Physical Form: Liquid
Density: 0.897
Appearance: Brown
Odor: Petroleum-like
UN No.: 1268
MDL No.: MFCD00058698
Formula:
C₁₆H₃₀MnO₄ Density: 0.897 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00058698 CAS Number: 13434-24-7 UN: 1268 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...