Cyclopentadienylmanganese tricarbonyl

Supplier: Thermo Scientific Chemicals
Danger

38606-03 38606-06
CAAA38606-03EA 92.59 CAD
CAAA38606-03 CAAA38606-06
Cyclopentadienylmanganese tricarbonyl
Cyclopentadienylmanganese tricarbonyl

CAS No.: 12079-65-1 Molecular Formula: C8H5MnO3 Formula Weight: 204.06 Physical Form: Crystalline Sensitivity: Air Sensitive Melting Point: 77° UN No.: 3466 MDL No.: MFCD00001440

Formula: C₈H₅MnO₃
Melting Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001440
CAS Number: 12079-65-1
EINECS: 235-142-4
UN: 3466
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR