Palladium 1% on activated carbon powder wetted with ca. 50% water, eggshell, reduced

Supplier: Thermo Scientific Chemicals

38527-06 38527-14
CAAA38527-06EA 77.58 CAD
CAAA38527-06 CAAA38527-14
Palladium 1% on activated carbon powder wetted with ca. 50% water, eggshell, reduced
Palladium

Sold on a dry weight basis. Unit weight excludes water weight


CAS No.: 7440-05-3 Physical Form: 900-1100m2/g MDL No.: MFCD03457879

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03457879
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR