Ruthenium 0.5% on carbon, granules
Supplier: Thermo Scientific Chemicals
38332-14
38332-22
CAAA38332-14EA
319.16
CAD
CAAA38332-14
CAAA38332-22
Ruthenium 0.5% on carbon, granules
Ruthenium
CAS No.: 7440-18-8
Physical Form: 4-12 Mesh, 900-1100m2/g
Odor: Odorless
UN No.: 1325
MDL No.: MFCD00011207
Formula:
Ru Storage Temperature: Ambient |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 EINECS: 231-127-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...