Platinum 10% on activated carbon wetted with ca. 50% water, powder, reduced

Supplier: Thermo Scientific Chemicals

38325-06 38325-14 38325-22
CAAA38325-06EA 178.54 CAD
CAAA38325-06 CAAA38325-14 CAAA38325-22
Platinum 10% on activated carbon wetted with ca. 50% water, powder, reduced
Platinum

Aromatic ring hydrogenation, aliphatic aldehydes and ketones to alcohols, conversion of -CH=CH- to -CH2-CH2-
Sold on a dry weight basis. Unit weight excludes water weight


CAS No.: 7440-06-4 Physical Form: 900-1100 m²/g Odor: Odorless MDL No.: MFCD00011179


Applications: Aromatic ring hydrogenation, aliphatic aldehydes and ketones to alcohols, conversion of -CH=CH- to -CH2-CH2-


Ordering information: Sold on a dry weight basis. Unit weight excludes water weight

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR