Platinum 3% on activated carbon wetted_50%_Water, powder, reduced
Supplier: Thermo Scientific Chemicals
Synonyms:
Platinium
38316-06
38316-14
CAAA38316-06EA
115.66
CAD
CAAA38316-06
CAAA38316-14
Platinum 3% on activated carbon wetted_50%_Water, powder, reduced
Platinum
Sold on a dry weight basis. Unit weight excludes water weight
CAS No.: 7440-06-4
Physical Form: 900-1100 m²/g
MDL No.: MFCD00011179
Ordering information: Sold on a dry weight basis. Unit weight excludes water weight
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...