Palladium 20% on activated carbon powder standard wetted with ca. 50% water, unreduced

Supplier: Thermo Scientific Chemicals

38309-06 38309-14 38309-22
CAAA38309-06EA 522.98 CAD
CAAA38309-06 CAAA38309-14 CAAA38309-22
Palladium 20% on activated carbon powder standard wetted with ca. 50% water, unreduced
Palladium

Physical Form: 900-1100 m²/g Odour: Odourless


Ordering information: Sold on a dry weight basis. Unit weight excludes water weight

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR