Palladium 20% on activated carbon powder standard wetted with ca. 50% water, reduced

Supplier: Thermo Scientific Chemicals

38308-04 38308-04 38308-09 38308-MD 38308-18 38308-09 38308-18 38308-MD
CAAA38308-18EA 4528.28 CAD
AA38308-09 CAAA38308-18 CAAA38308-MD CAAA38308-09 AA38308-MD AA38308-18 CAAA38308-04 AA38308-04
Palladium 20% on activated carbon powder standard wetted with ca. 50% water, reduced
Palladium

Ordering information: Sold on a dry weight basis. Unit weight excludes water weight

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR