Palladium 20% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced

Supplier: Thermo Scientific Chemicals

38307-06 38307-14
CAAA38307-06EA 522.98 CAD
CAAA38307-06 CAAA38307-14 CAAA38307-22
Palladium 20% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced
Palladium

Applications: Debenzylation
Sold on a dry weight basis. Unit weight excludes water weight


CAS No.: 7440-05-3 Physical Form: 900-1100m2/g Odor: Odorless MDL No.: MFCD03457879


Applications: Debenzylation

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR